Product Details

SNP ID
rs202231540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99982706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGGCGCAACTCCACAACCAGTA[C/T]GGCCCCATGAATATGAACATGGGTA
Phenotype
MIM: 603073
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZIC2 PubMed Links

Gene Details

Gene
ZIC2
Gene Name
Zic family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007129.3 706 Silent Mutation TAC,TAT Y214Y NP_009060.2
XM_011521110.2 706 Silent Mutation TAC,TAT Y214Y XP_011519412.1

View Full Product Details