Product Details

SNP ID
rs201966146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36432644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGGAGACCGGCTTTCCCGCAA[A/T]CATGTACCCTGGATCTTTTATTGGG
Phenotype
MIM: 604036
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNA1 PubMed Links

Gene Details

Gene
CCNA1
Gene Name
cyclin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111045.1 373 Missense Mutation AAC,ATC N8I NP_001104515.1
NM_001111046.1 373 Intron NP_001104516.1
NM_001111047.1 373 Intron NP_001104517.1
NM_003914.3 373 Missense Mutation AAC,ATC N8I NP_003905.1
XM_011535294.2 373 Intron XP_011533596.1
XM_011535295.2 373 Intron XP_011533597.1
XM_011535296.2 373 Intron XP_011533598.1

View Full Product Details