Product Details

SNP ID
rs201515700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113424301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCATAGGCTATCATGGGGGCAT[C/T]GTGGCCTCGTCTTCCCCCTCGACCT
Phenotype
MIM: 610620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADPRHL1 PubMed Links

Gene Details

Gene
ADPRHL1
Gene Name
ADP-ribosylhydrolase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138430.4 910 Missense Mutation AAT,GAT N275D NP_612439.2
NM_199162.2 910 Missense Mutation AAT,GAT N193D NP_954631.1
Gene
LOC101928841
Gene Name
collagen alpha-1(II) chain-like
There are no transcripts associated with this gene.

View Full Product Details