Product Details

SNP ID
rs201710720
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44989669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCACCGCGGATGGCAAGAGACCT[C/G]ATCGGACCGGCCCTGCCGCCCGGCT
Phenotype
MIM: 604354
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GPALPP1 PubMed Links

Gene Details

Gene
GPALPP1
Gene Name
GPALPP motifs containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316951.1 140 Silent Mutation CTC,CTG L5L NP_001303880.1
NM_001316952.1 140 UTR 5 NP_001303881.1
NM_018559.3 140 Silent Mutation CTC,CTG L5L NP_061029.2
XM_005266444.3 140 Silent Mutation CTC,CTG L5L XP_005266501.1
XM_011535144.2 140 UTR 5 XP_011533446.1
Gene
NUFIP1
Gene Name
NUFIP1, FMR1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012345.2 140 Intron NP_036477.2

View Full Product Details