Product Details

SNP ID
rs200424554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:96987222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGAGGTCGAGACAGGCTGATC[C/T]GTTGGTCCTGTTGGTTGATGTGATC
Phenotype
MIM: 606922
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00359 PubMed Links

Gene Details

Gene
LINC00359
Gene Name
long intergenic non-protein coding RNA 359
There are no transcripts associated with this gene.

Gene
OXGR1
Gene Name
oxoglutarate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080818.3 919 Missense Mutation AGA,GGA R180G NP_543008.3
XM_005254043.3 919 Missense Mutation AGA,GGA R180G XP_005254100.1
XM_005254044.3 919 Missense Mutation AGA,GGA R180G XP_005254101.1
XM_005254045.2 919 Missense Mutation AGA,GGA R180G XP_005254102.1
XM_005254046.2 919 Missense Mutation AGA,GGA R180G XP_005254103.1
XM_011521077.2 919 Missense Mutation AGA,GGA R180G XP_011519379.1

View Full Product Details