Product Details

SNP ID
rs200560214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27273186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTCCACCCTGACTCTAAAGCC[C/G]CTGTCATCATCGTGGGCAACAAGGG
Phenotype
MIM: 612403
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RASL11A PubMed Links

Gene Details

Gene
RASL11A
Gene Name
RAS like family 11 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206827.1 1039 Missense Mutation CCT,GCT P141A NP_996563.1

View Full Product Details