Product Details

SNP ID
rs199618630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39038707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCATTTTTGGTTTTGCATTCGC[G/T]TTTTTGTGGCTCTCCAGTGAGTTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NHLRC3 PubMed Links

Gene Details

Gene
NHLRC3
Gene Name
NHL repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012754.3 397 Missense Mutation CGT,CTT R23L NP_001012772.1
NM_001017370.2 397 Missense Mutation CGT,CTT R23L NP_001017370.1
Gene
PROSER1
Gene Name
proline and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025138.4 397 Intron NP_079414.3
XM_005266545.3 397 Intron XP_005266602.1
XM_011535239.2 397 Intron XP_011533541.1

View Full Product Details