Product Details
- SNP ID
-
rs200574898
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:113495714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGCGGGCGAGGGGTGGCTGCCATG[C/G]AGAGCCGGCAGTGGGTCCGGGACAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DCUN1D2
PubMed Links
Gene Details
- Gene
- DCUN1D2
- Gene Name
- defective in cullin neddylation 1 domain containing 2
There are no transcripts associated with this gene.
- Gene
- TMCO3
- Gene Name
- transmembrane and coiled-coil domains 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017905.4 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
NP_060375.4 |
XM_006719969.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_006720032.1 |
XM_011537498.2 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_011535800.1 |
XM_011537501.2 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_011535803.1 |
XM_011537502.2 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_011535804.1 |
XM_011537504.2 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_011535806.1 |
XM_011537506.2 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_011535808.1 |
XM_011537507.2 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_011535809.1 |
XM_011537509.2 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_011535811.1 |
XM_017020629.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876118.1 |
XM_017020630.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876119.1 |
XM_017020631.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876120.1 |
XM_017020632.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876121.1 |
XM_017020633.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876122.1 |
XM_017020634.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876123.1 |
XM_017020635.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876124.1 |
XM_017020636.1 |
494 |
Missense Mutation |
CAG,GAG |
Q45E |
XP_016876125.1 |
View Full Product Details