Product Details

SNP ID
rs200207534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:105472648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGGCATTTACAGAGATCATTA[C/T]GTCCTTGGGTCTCTTACCTTCCTCA
Phenotype
MIM: 607408 MIM: 607415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAOA PubMed Links

Gene Details

Gene
DAOA
Gene Name
D-amino acid oxidase activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161812.1 363 Silent Mutation TAC,TAT Y17Y NP_001155284.1
NM_001161814.1 363 Missense Mutation CGT,TGT R11C NP_001155286.1
NM_172370.4 363 Missense Mutation CGT,TGT R82C NP_758958.3
XM_005254042.1 363 Missense Mutation CGT,TGT R82C XP_005254099.1
Gene
DAOA-AS1
Gene Name
DAOA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details