Product Details
- SNP ID
-
rs200218010
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:33016916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTCGTGGGCGCGAGTGCTCCCCA[A/G]TGGCAGCGCGGGCGTCCCCAACCGC
- Phenotype
-
MIM: 604824
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KL
PubMed Links
Gene Details
- Gene
- KL
- Gene Name
- klotho
View Full Product Details