Product Details

SNP ID
rs200210579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21499048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGTTGGGTTGCACATTGTGTGG[C/T]CGTCCAAATAACTCAATCTTGCGAG
Phenotype
MIM: 612472 MIM: 614032
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METTL3 PubMed Links

Gene Details

Gene
METTL3
Gene Name
methyltransferase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019852.4 4342 Silent Mutation CGA,CGG R536R NP_062826.2
XM_006720206.3 4342 Silent Mutation CGA,CGG R252R XP_006720269.1
XM_011536968.1 4342 Silent Mutation CGA,CGG R514R XP_011535270.1
Gene
TOX4
Gene Name
TOX high mobility group box family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303523.1 4342 UTR 3 NP_001290452.1
NM_014828.3 4342 UTR 3 NP_055643.1

View Full Product Details