Product Details

SNP ID
rs200748907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77026318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTAGGGCTGGGGCAGCATGTCG[C/G]CGCCGGGCACGCCCTCCTTGAAGAA
Phenotype
MIM: 611720
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRF2BPL PubMed Links

Gene Details

Gene
IRF2BPL
Gene Name
interferon regulatory factor 2 binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024496.3 2382 Missense Mutation GCC,GGC A492G NP_078772.1
Gene
LOC105370579
Gene Name
uncharacterized LOC105370579
There are no transcripts associated with this gene.

Gene
LOC107984638
Gene Name
uncharacterized LOC107984638
There are no transcripts associated with this gene.

View Full Product Details