Product Details

SNP ID
rs201400690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23373216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGGTCATGGGAACCCACACCT[C/A]CAGCCACTGGCCCAGCTGCTGCCCC
Phenotype
MIM: 607888 MIM: 609906 MIM: 605658 MIM: 160710
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
CMTM5 PubMed Links

Gene Details

Gene
CMTM5
Gene Name
CKLF like MARVEL transmembrane domain containing 5
There are no transcripts associated with this gene.

Gene
EFS
Gene Name
embryonal Fyn-associated substrate
There are no transcripts associated with this gene.

Gene
IL25
Gene Name
interleukin 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022789.3 208 Missense Mutation TAC,TCC Y33S NP_073626.1
NM_172314.1 208 Missense Mutation TAC,TCC Y17S NP_758525.1
Gene
MYH6
Gene Name
myosin heavy chain 6
There are no transcripts associated with this gene.

View Full Product Details