Product Details
- SNP ID
-
rs201731132
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20289646 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAGGTCATTCACACTGTGGTCGC[A/G]ATGCCACTGTGGCAACAGCCTGGCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TTC5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3737220] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TTC5
- Gene Name
- tetratricopeptide repeat domain 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138376.2 |
1361 |
Missense Mutation |
TCG,TTG |
S435L |
NP_612385.2 |
View Full Product Details