Product Details

SNP ID
rs200002005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35534884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCACGGACTGCGCGTCTGGAC[A/C]CGCGGCCGCGGGAATCAAGAAGCCA
Phenotype
MIM: 614027 MIM: 608884
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INSM2 PubMed Links

Gene Details

Gene
INSM2
Gene Name
INSM transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032594.3 843 Missense Mutation CAC,CCC H211P NP_115983.3
Gene
RALGAPA1
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
There are no transcripts associated with this gene.

View Full Product Details