Product Details

SNP ID
rs200611630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22902006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAGAGCTGCTGGATTCAGGGCC[C/T]CCAAGGGAACTGCTCCATTCAGTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM23 PubMed Links

Gene Details

Gene
RBM23
Gene Name
RNA binding motif protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077351.1 1403 Missense Mutation GAG,GGG E407G NP_001070819.1
NM_001077352.1 1403 Missense Mutation GAG,GGG E373G NP_001070820.1
NM_001308044.1 1403 Missense Mutation GAG,GGG E237G NP_001294973.1
NM_018107.4 1403 Missense Mutation GAG,GGG E391G NP_060577.3
XM_011536890.2 1403 Missense Mutation GAG,GGG E451G XP_011535192.1
XM_011536892.2 1403 Missense Mutation GAG,GGG E435G XP_011535194.1
XM_011536893.2 1403 Missense Mutation GAG,GGG E435G XP_011535195.1
XM_011536894.2 1403 Missense Mutation GAG,GGG E433G XP_011535196.1
XM_011536895.2 1403 Missense Mutation GAG,GGG E417G XP_011535197.1
XM_011536896.2 1403 Missense Mutation GAG,GGG E417G XP_011535198.1
XM_011536897.2 1403 Missense Mutation GAG,GGG E409G XP_011535199.1
XM_011536900.1 1403 Missense Mutation GAG,GGG E389G XP_011535202.1
XM_011536902.1 1403 Missense Mutation GAG,GGG E237G XP_011535204.1
XM_011536903.1 1403 Missense Mutation GAG,GGG E237G XP_011535205.1
XM_011536904.1 1403 Missense Mutation GAG,GGG E237G XP_011535206.1
XM_011536905.1 1403 Missense Mutation GAG,GGG E237G XP_011535207.1
XM_011536906.1 1403 Missense Mutation GAG,GGG E237G XP_011535208.1
XM_017021398.1 1403 Missense Mutation GAG,GGG E435G XP_016876887.1
XM_017021399.1 1403 Missense Mutation GAG,GGG E417G XP_016876888.1
XM_017021400.1 1403 Missense Mutation GAG,GGG E409G XP_016876889.1
XM_017021401.1 1403 Missense Mutation GAG,GGG E407G XP_016876890.1
XM_017021402.1 1403 Missense Mutation GAG,GGG E407G XP_016876891.1
XM_017021403.1 1403 Missense Mutation GAG,GGG E391G XP_016876892.1
XM_017021404.1 1403 Missense Mutation GAG,GGG E391G XP_016876893.1
XM_017021405.1 1403 Missense Mutation GAG,GGG E389G XP_016876894.1
XM_017021406.1 1403 Missense Mutation GAG,GGG E425G XP_016876895.1
XM_017021407.1 1403 Missense Mutation GAG,GGG E407G XP_016876896.1
XM_017021408.1 1403 Missense Mutation GAG,GGG E373G XP_016876897.1
XM_017021409.1 1403 Missense Mutation GAG,GGG E407G XP_016876898.1
XM_017021410.1 1403 Missense Mutation GAG,GGG E373G XP_016876899.1
XM_017021411.1 1403 Missense Mutation GAG,GGG E391G XP_016876900.1
XM_017021412.1 1403 Intron XP_016876901.1
XM_017021413.1 1403 Intron XP_016876902.1
XM_017021414.1 1403 Missense Mutation GAG,GGG E237G XP_016876903.1
XM_017021415.1 1403 Missense Mutation GAG,GGG E237G XP_016876904.1
XM_017021416.1 1403 Missense Mutation GAG,GGG E237G XP_016876905.1
XM_017021417.1 1403 Missense Mutation GAG,GGG E237G XP_016876906.1
XM_017021418.1 1403 Missense Mutation GAG,GGG E237G XP_016876907.1
XM_017021419.1 1403 Missense Mutation GAG,GGG E237G XP_016876908.1
XM_017021420.1 1403 Missense Mutation GAG,GGG E237G XP_016876909.1

View Full Product Details