Product Details
- SNP ID
-
rs200830672
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20289714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCATTCACCACTAGCAGGAGGGG[C/T]GTCTCCACTCGAACACTGGAAAAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTC5
PubMed Links
Gene Details
- Gene
- TTC5
- Gene Name
- tetratricopeptide repeat domain 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138376.2 |
1293 |
Silent Mutation |
|
|
NP_612385.2 |
View Full Product Details