Product Details

SNP ID
rs201661122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73619277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTTCTTGGATTTTCCAAAGGAG[G/T]TGACCTGTGTCTCTCAATGGCTTCT
Phenotype
MIM: 614267
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACOT6 PubMed Links

Gene Details

Gene
ACOT6
Gene Name
acyl-CoA thioesterase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037162.1 393 Missense Mutation GGT,GTT G21V NP_001032239.1
XM_011537076.2 393 Missense Mutation GGT,GTT G273V XP_011535378.1
Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 393 Intron NP_001207413.1
NM_203309.2 393 Intron NP_976054.2
XM_006720143.2 393 Intron XP_006720206.1
XM_011536760.2 393 Intron XP_011535062.1
XM_011536761.2 393 Intron XP_011535063.1
XM_017021289.1 393 Intron XP_016876778.1

View Full Product Details