Product Details

SNP ID
rs200904933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75579404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTCCTGCCAGTGGCTTGGCT[G/T]CTGGAGAAGTTCGGCCTGCGCACCA
Phenotype
MIM: 610865
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FLVCR2 PubMed Links

Gene Details

Gene
FLVCR2
Gene Name
feline leukemia virus subgroup C cellular receptor family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195283.1 808 Intron NP_001182212.1
NM_017791.2 808 Silent Mutation CTG,CTT L144L NP_060261.2
Gene
LOC102724153
Gene Name
uncharacterized LOC102724153
There are no transcripts associated with this gene.

View Full Product Details