Product Details
- SNP ID
-
rs201159686
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:76154576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCCTCTGAGTCAAGTCTGGATGA[C/G]GCCACTAAGGACTGTCGAGAAGTGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPATCH2L
PubMed Links
Gene Details
- Gene
- GPATCH2L
- Gene Name
- G-patch domain containing 2 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322026.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_001308955.1 |
NM_001322027.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_001308956.1 |
NM_001322028.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_001308957.1 |
NM_001322029.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_001308958.1 |
NM_001322030.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_001308959.1 |
NM_001322031.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_001308960.1 |
NM_001322032.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_001308961.1 |
NM_017926.3 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_060396.2 |
NM_017972.3 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
NP_060442.2 |
XM_006720191.2 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
XP_006720254.1 |
XM_017021427.1 |
530 |
Missense Mutation |
GAC,GAG |
D84E |
XP_016876916.1 |
XM_017021428.1 |
530 |
Missense Mutation |
GAC,GAG |
D84E |
XP_016876917.1 |
XM_017021429.1 |
530 |
Missense Mutation |
GAC,GAG |
D84E |
XP_016876918.1 |
XM_017021430.1 |
530 |
Missense Mutation |
GAC,GAG |
D84E |
XP_016876919.1 |
XM_017021431.1 |
530 |
Missense Mutation |
GAC,GAG |
D84E |
XP_016876920.1 |
XM_017021432.1 |
530 |
Missense Mutation |
GAC,GAG |
D84E |
XP_016876921.1 |
XM_017021433.1 |
530 |
Missense Mutation |
GAC,GAG |
D71E |
XP_016876922.1 |
View Full Product Details