Product Details
- SNP ID
-
rs201193955
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:68877194 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCTGGAATGTCACTACCCCCAGG[C/T]GGTTGGGGTCCACAATGCTCATGAT
- Phenotype
-
MIM: 102575
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTN1
PubMed Links
Gene Details
- Gene
- ACTN1
- Gene Name
- actinin alpha 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001102.3 |
3360 |
Missense Mutation |
CAC,CGC |
H803R |
NP_001093.1 |
NM_001130004.1 |
3360 |
Missense Mutation |
CAC,CGC |
H825R |
NP_001123476.1 |
NM_001130005.1 |
3360 |
Missense Mutation |
CAC,CGC |
H798R |
NP_001123477.1 |
XM_011537265.2 |
3360 |
Missense Mutation |
CAC,CGC |
H840R |
XP_011535567.1 |
XM_011537266.2 |
3360 |
Missense Mutation |
CAC,CGC |
H835R |
XP_011535568.1 |
XM_011537267.2 |
3360 |
Missense Mutation |
CAC,CGC |
H832R |
XP_011535569.1 |
XM_011537268.2 |
3360 |
Missense Mutation |
CAC,CGC |
H827R |
XP_011535570.1 |
XM_017021720.1 |
3360 |
Missense Mutation |
CAC,CGC |
H1017R |
XP_016877209.1 |
XM_017021721.1 |
3360 |
Missense Mutation |
CAC,CGC |
H1009R |
XP_016877210.1 |
XM_017021722.1 |
3360 |
Missense Mutation |
CAC,CGC |
H988R |
XP_016877211.1 |
XM_017021723.1 |
3360 |
Missense Mutation |
CAC,CGC |
H980R |
XP_016877212.1 |
XM_017021725.1 |
3360 |
Missense Mutation |
CAC,CGC |
H967R |
XP_016877214.1 |
XM_017021726.1 |
3360 |
Missense Mutation |
CAC,CGC |
H959R |
XP_016877215.1 |
XM_017021727.1 |
3360 |
Missense Mutation |
CAC,CGC |
H806R |
XP_016877216.1 |
XM_017021728.1 |
3360 |
Missense Mutation |
CAC,CGC |
H777R |
XP_016877217.1 |
View Full Product Details