Product Details

SNP ID
rs200871983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24336244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGGATCTTTAGGGCCTTCTTG[C/T]GAACCTACTGGTGGGGGGTGCTGCA
Phenotype
MIM: 600292 MIM: 605817
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCY4 PubMed Links

Gene Details

Gene
ADCY4
Gene Name
adenylate cyclase 4
There are no transcripts associated with this gene.

Gene
RIPK3
Gene Name
receptor interacting serine/threonine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006871.3 1698 Silent Mutation TCA,TCG S496S NP_006862.2

View Full Product Details