Product Details

SNP ID
rs201198198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99174214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCAAGTGCTCGCCGTGCCACTT[C/T]TTCATGTGTTTCTCCAGGGTGCTGT
Phenotype
MIM: 606558
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL11B PubMed Links

Gene Details

Gene
BCL11B
Gene Name
B-cell CLL/lymphoma 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282237.1 2904 Silent Mutation AAA,AAG K873K NP_001269166.1
NM_001282238.1 2904 Silent Mutation AAA,AAG K802K NP_001269167.1
NM_022898.2 2904 Silent Mutation AAA,AAG K803K NP_075049.1
NM_138576.3 2904 Silent Mutation AAA,AAG K874K NP_612808.1

View Full Product Details