Product Details

SNP ID
rs201162952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:56580009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCATCCGCGGCGGCGTGGCGG[T/G]GTTCGCCGCCGTGGCCGCAGTGTTC
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM260 PubMed Links

Gene Details

Gene
TMEM260
Gene Name
transmembrane protein 260
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017799.3 184 Missense Mutation GGG,GTG G32V NP_060269.3
XM_005267771.1 184 Intron XP_005267828.1
XM_006720176.1 184 UTR 5 XP_006720239.1
XM_006720178.1 184 UTR 5 XP_006720241.1
XM_011536851.1 184 Missense Mutation GGG,GTG G32V XP_011535153.1
XM_011536852.1 184 UTR 5 XP_011535154.1
XM_011536853.1 184 UTR 5 XP_011535155.1
XM_017021379.1 184 Missense Mutation GGG,GTG G32V XP_016876868.1
XM_017021380.1 184 UTR 5 XP_016876869.1

View Full Product Details