Product Details

SNP ID
rs200159534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73492589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGGAGAGGGCACTAAGTGTTTA[C/T]GGGCTTCCAATTCGCTGGGAGGCTG
Phenotype
MIM: 614313 MIM: 611919
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT1 PubMed Links

Gene Details

Gene
ACOT1
Gene Name
acyl-CoA thioesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037161.1 1656 Intron NP_001032238.1
XM_017021589.1 1656 Intron XP_016877078.1
XM_017021590.1 1656 Intron XP_016877079.1
XM_017021591.1 1656 Intron XP_016877080.1
XM_017021592.1 1656 Intron XP_016877081.1
Gene
C14orf169
Gene Name
chromosome 14 open reading frame 169
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024644.4 1656 Silent Mutation TAC,TAT Y524Y NP_078920.2
Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 1656 Intron NP_001207413.1
NM_203309.2 1656 Intron NP_976054.2
XM_006720143.2 1656 Intron XP_006720206.1
XM_011536760.2 1656 Intron XP_011535062.1
XM_011536761.2 1656 Intron XP_011535063.1
XM_017021289.1 1656 Intron XP_016876778.1

View Full Product Details