Product Details

SNP ID
rs201561302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20117891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTGGCCATTTGTAAGCCCCTAC[A/G]CTACATGACCATCATGAACAAGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4K17 PubMed Links

Gene Details

Gene
OR4K17
Gene Name
olfactory receptor family 4 subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004715.1 485 Missense Mutation CAC,CGC H162R NP_001004715.1

View Full Product Details