Product Details

SNP ID
rs200409369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105173081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACCATGAGACACCAAAGATGAC[A/G]CCGAGGGTCTTGGGGATGTCCGGGT
Phenotype
MIM: 602570 MIM: 609219
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JAG2 PubMed Links

Gene Details

Gene
JAG2
Gene Name
jagged 2
There are no transcripts associated with this gene.

Gene
NUDT14
Gene Name
nudix hydrolase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318380.1 723 Intron NP_001305309.1
NM_177533.4 723 Silent Mutation GGC,GGT G203G NP_803877.2

View Full Product Details