Product Details

SNP ID
rs201889259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103532814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACATCCCATCACCCTGGGAGGC[C/T]GTGGGCCACGCCTGGGACGCCCTCA
Phenotype
MIM: 123280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CKB PubMed Links

Gene Details

Gene
CKB
Gene Name
creatine kinase B
There are no transcripts associated with this gene.

Gene
TRMT61A
Gene Name
tRNA methyltransferase 61A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152307.2 683 Silent Mutation GCC,GCT A188A NP_689520.2

View Full Product Details