Product Details

SNP ID
rs200163818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77278484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCCAAGCCCCAGGCACAGAGC[C/T]GCAGGAGGGTGTCCCACAGAATGCC
Phenotype
MIM: 605304 MIM: 607439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NGB PubMed Links

Gene Details

Gene
NGB
Gene Name
neuroglobin
There are no transcripts associated with this gene.

Gene
POMT2
Gene Name
protein O-mannosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013382.5 2360 Missense Mutation CAG,CGG Q686R NP_037514.2
XM_011536675.2 2360 Missense Mutation CAG,CGG Q749R XP_011534977.1
XM_011536676.2 2360 Missense Mutation CAG,CGG Q638R XP_011534978.1
XM_011536677.2 2360 Missense Mutation CAG,CGG Q596R XP_011534979.1
XM_011536679.2 2360 Missense Mutation CAG,CGG Q447R XP_011534981.1
XM_017021244.1 2360 Missense Mutation CAG,CGG Q384R XP_016876733.1
XM_017021245.1 2360 Missense Mutation CAG,CGG Q384R XP_016876734.1

View Full Product Details