Product Details

SNP ID
rs200342853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75525104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGACTCATCTGATGATGTGAG[A/G]AGAGTTCAGAGGAGGGAGAAAAATC
Phenotype
MIM: 612476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BATF PubMed Links

Gene Details

Gene
BATF
Gene Name
basic leucine zipper ATF-like transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006399.3 326 Silent Mutation AGA,AGG R28R NP_006390.1

View Full Product Details