Product Details

SNP ID
rs201603417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23048355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGCCGTGATGTCGAAGGCCTC[G/T]GTGTCCTCCTCGCCGCCGCCCTCGT
Phenotype
MIM: 611137
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDH24 PubMed Links

Gene Details

Gene
CDH24
Gene Name
cadherin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022478.3 2345 Silent Mutation NP_071923.2
NM_144985.3 2345 Silent Mutation NP_659422.2
XM_011537089.1 2345 Intron XP_011535391.1
Gene
LOC105370705
Gene Name
collagen alpha-2(I) chain-like
There are no transcripts associated with this gene.

Gene
PSMB11
Gene Name
proteasome subunit beta 11
There are no transcripts associated with this gene.

View Full Product Details