Product Details

SNP ID
rs200810211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103530113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCGGCATGGTGTCCTGCGGCA[C/T]TCAGTTGACCTTATCGGCCGCCCCT
Phenotype
MIM: 123280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CKB PubMed Links

Gene Details

Gene
CKB
Gene Name
creatine kinase B
There are no transcripts associated with this gene.

Gene
TRMT61A
Gene Name
tRNA methyltransferase 61A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152307.2 254 Silent Mutation CAC,CAT H45H NP_689520.2

View Full Product Details