Product Details

SNP ID
rs201355660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52315218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGTGTCATTCTCAACCTCATC[C/G]GCATGCACCGCCGAAGCCGGAGAAG
Phenotype
MIM: 176804
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTGER2 PubMed Links

Gene Details

Gene
PTGER2
Gene Name
prostaglandin E receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000956.3 921 Missense Mutation CGC,GGC R224G NP_000947.2

View Full Product Details