Product Details
- SNP ID
-
rs200992969
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:57471378 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTATTCTTGATCAAAAAACTCAT[C/T]GAAAAGTGGGATTCTCTCTGTCCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C14orf105
PubMed Links
Gene Details
- Gene
- C14orf105
- Gene Name
- chromosome 14 open reading frame 105
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001283056.1 |
1136 |
Missense Mutation |
AAT,GAT |
N330D |
NP_001269985.1 |
NM_001283057.1 |
1136 |
Missense Mutation |
AAT,GAT |
N289D |
NP_001269986.1 |
NM_001283058.1 |
1136 |
Missense Mutation |
AAT,GAT |
N201D |
NP_001269987.1 |
NM_001283059.1 |
1136 |
Missense Mutation |
AAT,GAT |
N201D |
NP_001269988.1 |
NM_001283060.1 |
1136 |
Intron |
|
|
NP_001269989.1 |
NM_018168.3 |
1136 |
Missense Mutation |
AAT,GAT |
N290D |
NP_060638.2 |
XM_005267806.1 |
1136 |
Intron |
|
|
XP_005267863.1 |
XM_005267810.3 |
1136 |
Missense Mutation |
AAT,GAT |
N211D |
XP_005267867.1 |
XM_005267811.3 |
1136 |
Missense Mutation |
AAT,GAT |
N202D |
XP_005267868.1 |
XM_005267813.1 |
1136 |
Missense Mutation |
AAT,GAT |
N164D |
XP_005267870.1 |
XM_005267814.1 |
1136 |
Missense Mutation |
AAT,GAT |
N163D |
XP_005267871.1 |
XM_006720188.2 |
1136 |
Intron |
|
|
XP_006720251.1 |
XM_006720189.2 |
1136 |
Missense Mutation |
AAT,GAT |
N212D |
XP_006720252.1 |
View Full Product Details