Product Details
- SNP ID
-
rs201028153
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:73595265 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTTTCTCAGGCCTCGTGGACATT[A/G]TGGATATAAGGAATGCTCTCGTAGG
- Phenotype
-
MIM: 614314
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACOT4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2010070] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACOT4
- Gene Name
- acyl-CoA thioesterase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152331.3 |
|
Intron |
|
|
NP_689544.3 |
- Gene
- HEATR4
- Gene Name
- HEAT repeat containing 4
View Full Product Details