Product Details

SNP ID
rs201392957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:62707819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCAGCCTGGATGGGACTGTGCT[C/T]TAGCGGACTTCGGGCCTCCCCAGCC
Phenotype
MIM: 605716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNH5 PubMed Links

Gene Details

Gene
KCNH5
Gene Name
potassium voltage-gated channel subfamily H member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139318.4 2511 Missense Mutation AAG,GAG K886E NP_647479.2
NM_172375.2 2511 UTR 3 NP_758963.1

View Full Product Details