Product Details
- SNP ID
-
rs199880599
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:21102110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCTGGAGCTTCGCAGAGCACCCC[A/G]CCTTTGGAAGGTGAGAGGGAAGAAA
- Phenotype
-
MIM: 605036
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM253
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs745696] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM253
- Gene Name
- transmembrane protein 253
- Gene
- ZNF219
- Gene Name
- zinc finger protein 219
View Full Product Details