Product Details

SNP ID
rs200349818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74946266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTCCTCTTCCCCCTGCCCTT[A/G]GGTCTCCTCCTGCAATAAGCCAAGC
Phenotype
MIM: 601121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGF PubMed Links

Gene Details

Gene
PGF
Gene Name
placental growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207012.1 954 Intron NP_001193941.1
NM_001293643.1 954 Silent Mutation CCC,CCT P143P NP_001280572.1
NM_002632.5 954 Silent Mutation CCC,CCT P144P NP_002623.2

View Full Product Details