Product Details
- SNP ID
-
rs201501111
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:92797487 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAAGGCAGAAGATCTTTTAAACC[A/G]AGTTGATCAAGGGGCTGCAACAGCT
- Phenotype
-
MIM: 606918
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGA5
PubMed Links
Gene Details
- Gene
- GOLGA5
- Gene Name
- golgin A5
View Full Product Details