Product Details

SNP ID
rs199847263
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59472986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTCACTTGAGAAGAAATCCATC[A/C]CTCAATGGGTCGTCATCTTCTATTA
Phenotype
MIM: 607970 MIM: 614811
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GPR135 PubMed Links

Gene Details

Gene
GPR135
Gene Name
G protein-coupled receptor 135
There are no transcripts associated with this gene.

Gene
L3HYPDH
Gene Name
trans-L-3-hydroxyproline dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144581.1 2602 Missense Mutation AGG,AGT R348S NP_653182.1
XM_005267298.3 2602 Missense Mutation AGG,AGT R177S XP_005267355.1
XM_011536386.2 2602 Intron XP_011534688.1
XM_011536387.1 2602 UTR 3 XP_011534689.1
XM_011536388.1 2602 UTR 3 XP_011534690.1

View Full Product Details