Product Details

SNP ID
rs199883538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20640948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATAGAAGAGGGTCACCACTAT[C/G]AAGTGGGAGGTACAGGTAGAGAAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR6S1 PubMed Links

Gene Details

Gene
OR6S1
Gene Name
olfactory receptor family 6 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001968.1 744 Missense Mutation TTC,TTG F248L NP_001001968.1

View Full Product Details