Product Details

SNP ID
rs200421984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70459595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGTCTGTTTTGACTTTCCTGT[C/T]TACTGTCGGGCTTCTTATGTATCTA
Phenotype
MIM: 603713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM20P1 PubMed Links

Gene Details

Gene
ADAM20P1
Gene Name
ADAM metallopeptidase domain 20 pseudogene 1
There are no transcripts associated with this gene.

Gene
ADAM21
Gene Name
ADAM metallopeptidase domain 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003813.3 2354 Missense Mutation TCT,TTT S699F NP_003804.2

View Full Product Details