Product Details

SNP ID
rs200818825
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23275611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCACATCATCATCATCATCATC[A/T]TCATCTTCCTCCTCCTCCTCCTCCT
Phenotype
MIM: 608119
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
HOMEZ PubMed Links

Gene Details

Gene
HOMEZ
Gene Name
homeobox and leucine zipper encoding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020834.2 1765 Missense Mutation GAA,GAT E539D NP_065885.2
Gene
RNF212B
Gene Name
ring finger protein 212B
There are no transcripts associated with this gene.

View Full Product Details