Product Details

SNP ID
rs202181054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22974846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCCAGACATACTGACCTCAGAG[A/G]GGAGGATGGGTTGGCCACAGGCTGC
Phenotype
MIM: 609066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AJUBA PubMed Links

Gene Details

Gene
AJUBA
Gene Name
ajuba LIM protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289097.1 1791 Intron NP_001276026.1
NM_032876.5 1791 Missense Mutation CCC,CTC P472L NP_116265.1
NM_198086.2 1791 Missense Mutation CCC,CTC P55L NP_932352.1

View Full Product Details