Product Details

SNP ID
rs201968492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:67340460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACCTATAGAACTCTTCTCGCTC[G/T]CTCTCATCCAGCTCTGTGATGATAT
Phenotype
MIM: 609398 MIM: 603930 MIM: 606958
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1D PubMed Links

Gene Details

Gene
ATP6V1D
Gene Name
ATPase H+ transporting V1 subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015994.3 887 Missense Mutation AGA,AGC R194S NP_057078.1
Gene
GPHN
Gene Name
gephyrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024218.1 887 Intron NP_001019389.1
NM_020806.4 887 Intron NP_065857.1
XM_005267254.3 887 Intron XP_005267311.1
XM_011536340.2 887 Intron XP_011534642.1
XM_011536342.2 887 Intron XP_011534644.1
XM_011536343.2 887 Intron XP_011534645.1
XM_011536344.2 887 Intron XP_011534646.1
XM_011536345.2 887 Intron XP_011534647.1
XM_011536346.2 887 Intron XP_011534648.1
XM_011536347.2 887 Intron XP_011534649.1
XM_017020913.1 887 Intron XP_016876402.1
XM_017020914.1 887 Intron XP_016876403.1
XM_017020915.1 887 Intron XP_016876404.1
XM_017020916.1 887 Intron XP_016876405.1
XM_017020917.1 887 Intron XP_016876406.1
XM_017020918.1 887 Intron XP_016876407.1
XM_017020919.1 887 Intron XP_016876408.1
XM_017020920.1 887 Intron XP_016876409.1
XM_017020921.1 887 Intron XP_016876410.1
XM_017020922.1 887 Intron XP_016876411.1
XM_017020923.1 887 Intron XP_016876412.1
XM_017020924.1 887 Intron XP_016876413.1
XM_017020925.1 887 Intron XP_016876414.1
XM_017020926.1 887 Intron XP_016876415.1
Gene
LOC105370541
Gene Name
uncharacterized LOC105370541
There are no transcripts associated with this gene.

Gene
MPP5
Gene Name
membrane palmitoylated protein 5
There are no transcripts associated with this gene.

View Full Product Details