Product Details
- SNP ID
-
rs202044227
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:74909108 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCTCCCCCCTTCTTGCCTGGGGC[A/G]CTGTAGAGATTGTCCACGGCCTCCC
- Phenotype
-
MIM: 126063
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DLST
PubMed Links
Gene Details
- Gene
- DLST
- Gene Name
- dihydrolipoamide S-succinyltransferase
There are no transcripts associated with this gene.
- Gene
- RPS6KL1
- Gene Name
- ribosomal protein S6 kinase like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031464.4 |
1476 |
Silent Mutation |
AGA,AGG |
R451R |
NP_113652.2 |
XM_011537206.2 |
1476 |
Silent Mutation |
AGA,AGG |
R467R |
XP_011535508.1 |
XM_011537207.2 |
1476 |
Silent Mutation |
AGA,AGG |
R467R |
XP_011535509.1 |
XM_011537211.2 |
1476 |
Silent Mutation |
AGA,AGG |
R420R |
XP_011535513.1 |
XM_011537213.2 |
1476 |
Silent Mutation |
AGA,AGG |
R404R |
XP_011535515.1 |
XM_017021680.1 |
1476 |
Silent Mutation |
AGA,AGG |
R467R |
XP_016877169.1 |
XM_017021681.1 |
1476 |
Silent Mutation |
AGA,AGG |
R451R |
XP_016877170.1 |
XM_017021682.1 |
1476 |
Missense Mutation |
AGA,AGG |
R451R |
XP_016877171.1 |
XM_017021683.1 |
1476 |
Intron |
|
|
XP_016877172.1 |
XM_017021684.1 |
1476 |
Silent Mutation |
AGA,AGG |
R467R |
XP_016877173.1 |
XM_017021685.1 |
1476 |
Silent Mutation |
AGA,AGG |
R467R |
XP_016877174.1 |
XM_017021686.1 |
1476 |
Missense Mutation |
AGC,GGC |
S458G |
XP_016877175.1 |
XM_017021687.1 |
1476 |
Missense Mutation |
AGC,GGC |
S458G |
XP_016877176.1 |
XM_017021688.1 |
1476 |
Silent Mutation |
AGA,AGG |
R265R |
XP_016877177.1 |
XM_017021689.1 |
1476 |
Silent Mutation |
AGA,AGG |
R265R |
XP_016877178.1 |
XM_017021690.1 |
1476 |
Silent Mutation |
AGA,AGG |
R249R |
XP_016877179.1 |
XM_017021691.1 |
1476 |
Intron |
|
|
XP_016877180.1 |
XM_017021692.1 |
1476 |
Intron |
|
|
XP_016877181.1 |
View Full Product Details