Product Details

SNP ID
rs202157686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:28768426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCACGCACCACCTCACGGCCGCC[A/G]CGCTAGCCGCCTCGGTGCCCTGCGG
Phenotype
MIM: 164874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXG1 PubMed Links

Gene Details

Gene
FOXG1
Gene Name
forkhead box G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005249.4 1355 Missense Mutation ACG,GCG T383A NP_005240.3
Gene
FOXG1-AS1
Gene Name
FOXG1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
LINC01551
Gene Name
long intergenic non-protein coding RNA 1551
There are no transcripts associated with this gene.

View Full Product Details