Product Details

SNP ID
rs201989906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52005771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGGGGTAGACTGCAGTTATCCC[A/G]TAGAGGTGAGATCGTTGTTCTGGGA
Phenotype
MIM: 610858 MIM: 605399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf166 PubMed Links

Gene Details

Gene
C14orf166
Gene Name
chromosome 14 open reading frame 166
There are no transcripts associated with this gene.

Gene
NID2
Gene Name
nidogen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007361.3 4317 Silent Mutation TAC,TAT Y1361Y NP_031387.3
XM_005267405.4 4317 Silent Mutation TAC,TAT Y1388Y XP_005267462.1
XM_005267406.4 4317 Silent Mutation TAC,TAT Y1340Y XP_005267463.1
XM_005267407.4 4317 Silent Mutation TAC,TAT Y1313Y XP_005267464.1

View Full Product Details