Product Details

SNP ID
rs202029589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20060138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATTTTCTTCTTTGTGACATTT[G/T]CCCTGATCTACGGTGCTACTGTGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4L1 PubMed Links

Gene Details

Gene
OR4L1
Gene Name
olfactory receptor family 4 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004717.1 94 Missense Mutation GCC,TCC A32S NP_001004717.1

View Full Product Details