Product Details

SNP ID
rs193920797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100325815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGCGGTGTTGCTGTGGGCTACC[C/T]CCTGGACACGGTGAAGGTGCGGCAA
Phenotype
MIM: 609911 MIM: 191050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A47 PubMed Links

Gene Details

Gene
SLC25A47
Gene Name
solute carrier family 25 member 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207117.2 129 Missense Mutation CCC,CTC P19L NP_997000.2
Gene
WARS
Gene Name
tryptophanyl-tRNA synthetase
There are no transcripts associated with this gene.

View Full Product Details